missing translation for 'onlineSavingsMsg'
Learn More

Bromotrimethylsilane, 98%

Product Code. 11915811 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11915811 500 g Glass bottle 500g
10276000 5 g Glass bottle 5g
10347940 25 g Glass bottle 25g
10052300 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11915811 Supplier Thermo Scientific Acros Supplier No. 186655000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2857-97-8 | C3H9BrSi | 153.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Silylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2857-97-8
Molecular Formula C3H9BrSi
Molecular Weight (g/mol) 153.1
InChI Key IYYIVELXUANFED-UHFFFAOYSA-N
Synonym trimethylsilyl bromide, trimethylbromosilane, silane, bromotrimethyl, trimethylsilicon bromide, tmsbr, trimethylsilylbromide, bromotrimethyl silane, tmbs, bromo trimethyl silane, tms bromide
PubChem CID 76113
IUPAC Name bromo(trimethyl)silane
SMILES C[Si](C)(C)Br

Specifications

Melting Point -43.0°C
Color Colorless to Yellow
Density 1.1600g/mL
Boiling Point 79.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (CH3)3SiBr
Refractive Index 1.4200 to 1.4250
Quantity 500 g
Fieser 09,73; 10,59; 12,79; 15,51; 16,50
Specific Gravity 1.16
Solubility Information Solubility in water: decomposes.
Formula Weight 153.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Bromotrimethylsilane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.