missing translation for 'onlineSavingsMsg'
Learn More

Bronopol, TRC

Product Code. 30340396
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30340396 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340396 Supplier Toronto Research Chemicals Supplier No. TRCB80990350G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bronopol
CAS 52-51-7
Molecular Formula C3 H6 Br N O4
Quantity 50 g
Synonym 2-Bromo-2-nitro-1,3-propanediol, 2-Bromo-2-nitropropan-1,3-diol, 2-Bromo-2-nitropropane-1,3-diol, 2-Nitro-2-bromo-1,3-propanediol, Acticide L 30, BE 6, BE 6 (bactericide), BNPD, BNPK, Bactrinashak, Bactrinol 100, Bactronol, Bioban, Bioban BP Plus, Bronidiol, Bronocot, Bronopol, Bronotak, Busan 1144, Canguard 409, Myacide AS, Myacide AS Plus, Myacide BT, Myacide Pharma BP, N 25, N 25 (antimicrobial), NSC 141021, Nalco 92RU093, Onyxide 500, Preventol P 100, Protectol BN 98, Protectol BN 99, Pyceze, Topcide 2520, Ultra-Fresh SAB
InChI Formula InChI=1S/C3H6BrNO4/c4-3(1-6,2-7)5(8)9/h6-7H,1-2H2
SMILES OCC(Br)(CO)[N+](=O)[O-]
IUPAC Name 2-bromo-2-nitropropane-1,3-diol
Molecular Weight (g/mol) 199.99
Formula Weight 198.948
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.