missing translation for 'onlineSavingsMsg'
Learn More

Brucine, 99%, anhydrous

Product Code. 10227773 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10227773 5 g Glass bottle 5g
10411681 50 g Glass bottle 50g
10675511 100 g Glass bottle 100g
10297303 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10227773 Supplier Thermo Scientific Acros Supplier No. 278720050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 357-57-3 | C23H26N2O4 | 394.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 357-57-3
Molecular Formula C23H26N2O4
Molecular Weight (g/mol) 394.46
MDL Number MFCD00005942
InChI Key RRKTZKIUPZVBMF-VLTSECPOSA-N
Synonym brucine, anhydrous, brucine:strychnidin-10-one,2,3-dimethoxy
PubChem CID 51413923
IUPAC Name (4aR,5aS,8aR,13aS,15aR,15bS)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one
SMILES COC1=C(C=C2C(=C1)C34CCN5C3CC6C7C4N2C(=O)CC7OCC=C6C5)OC

Specifications

Melting Point 174.0°C to 179.0°C
Color White to Beige
Loss on Drying 1% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Beilstein 27, IV, 7876
Merck Index 15, 1464
Specific Rotation Condition −123° (20°C c=2.5,CCl3)
Specific Rotation − 123.00
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, chloroform and benzene, slightly soluble in ether, glycerol and, ethyl acetate
Formula Weight 394.46
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Brucine, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.