missing translation for 'onlineSavingsMsg'
Learn More

Brucine dihydrate, 99%

Product Code. 10503531 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10503531 50 g Glass bottle 50g
10782391 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10503531 Supplier Thermo Scientific Acros Supplier No. 163500500
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 145428-94-0 | C23H27N2O4 | 395.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145428-94-0
Molecular Formula C23H27N2O4
Molecular Weight (g/mol) 395.48
MDL Number MFCD00005942
InChI Key RRKTZKIUPZVBMF-IBTVXLQLSA-O
Synonym brucine dihydrate, 10,11-dimethoxystrychnine, 10,11-dimethoxystrychnine dihydrate
PubChem CID 69897601
IUPAC Name (4aR,5aS,8aR,13aS,15aS,15bR)-10,11-dimethoxy-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;dihydrate
SMILES COC1=CC2=C(C=C1OC)[C@@]13CC[NH+]4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34

Specifications

Melting Point 174°C to 179°C
Color White to Beige
Loss on Drying 6.0% to 9.5% (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 50 g
Merck Index 14, 1455
Specific Rotation -123
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,chloroform and benzene,slightly soluble in ether,glycerol and,ethyl acetate
Formula Weight 430.5
Percent Purity 99%
Physical Form Crystals or Powder
Chemical Name or Material Brucine dihydrate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Brucine dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H330-Fatal if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.