Learn More
Paragon Scientific™ BS/IP/SL(S) Viscometer for Transparent Liquids (UKAS ISO 17025 certified)
Product Code. 16090380
Change view
Click to view available options
Viscosity Range:
1.05 min
2.1 to 3 cSt
20 to 100 cSt
200 to 1000 cSt
2000 to 10,000 cSt
3.8 to 10 cSt
6 to 30 cSt
60 to 300 cSt
600 to 3000 cSt
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity Range unitSize
16090380 1.05 min Each
16010390 2.1 to 3 cSt Each
16030390 3.8 to 10 cSt Each
16050390 6 to 30 cSt Each
16070390 20 to 100 cSt Each
16090390 60 to 300 cSt Each
16010400 200 to 1000 cSt Each
16030400 600 to 3000 cSt Each
16050400 2000 to 10,000 cSt Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 16090380 Supplier Paragon Scientific™ Supplier No. SHORT01UKC
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For the determination of kinematic viscosity of transparent Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
  • Supplied with full UKAS ISO 17025 calibration
Specifications
Viscosity Range 1.05 min
Calibrated Yes
Approximate Constant 0.0008
ASTM Numbers D 445, D 446
Certifications/Compliance ASTM D 445, D 446 and ISO 3104, 3105
Includes UKAS ISO 17025 calibration
Size 1
8 week lead-time