Learn More
Paragon Scientific™ BS/IP/SL Viscometer for Transparent Liquids (non certified)
Product Code. 15969939
Change view
Click to view available options
Viscosity Range:
20 to 100 cSt
20,000 to 100,000 cSt
200 to 1000 cSt
2000 to 10,000 cSt
3.5 to 10 cSt
6 to 30 cSt
60 to 300 cSt
600 to 3000 cSt
6000 to 30,000 cSt
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity Range unitSize
15969939 3.5 to 10 cSt Each
15949939 6 to 30 cSt Each
15909949 20 to 100 cSt Each
15989939 60 to 300 cSt Each
15949949 200 to 1000 cSt Each
15929949 600 to 3000 cSt Each
15989949 2000 to 10,000 cSt Each
15969949 6000 to 30,000 cSt Each
15909959 20,000 to 100,000 cSt Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15969939 Supplier Paragon Scientific™ Supplier No. BSIPSL1NC
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For the determination of kinematic viscosity of transparent Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
Viscosity Range 3.5 to 10 cSt
Calibrated No
Approximate Constant 0.01
ASTM Numbers D 445, D 446
Certifications/Compliance ASTM D 445, D 446 and ISO 3104, 3105
Size 1
4 week lead-time