Learn More
Paragon Scientific™ BS/U-tube Viscometer for Transparent Liquids
Product Code. 16060460
Change view
Click to view available options
Viscosity Range:
0.9 to 3 cSt
2.0 to 10 cSt
20 to 100 cSt
200 to 1000 cSt
2000 to 10,000 cSt
6 to 30 cSt
60 to 300 cSt
600 to 3000 cSt
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity Range unitSize
16060460 60 to 300 cSt Each
16080450 0.9 to 3 cSt Each
16000460 2.0 to 10 cSt Each
16020460 6 to 30 cSt Each
16040460 20 to 100 cSt Each
16080460 200 to 1000 cSt Each
16000470 600 to 3000 cSt Each
16020470 2000 to 10,000 cSt Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 16060460 Supplier Paragon Scientific™ Supplier No. U05NC
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For the determination of kinematic viscosity of transparent Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
Viscosity Range 60 to 300 cSt
Calibrated No
Approximate Constant 0.3
ASTM Numbers D 445, D 446
Certifications/Compliance ASTM D 445, D 446 and ISO 3104, 3105
Size E
4 week lead-time