Learn More
BTX™ Gemini X2 Electroporation Generator Footswitch
Product Code. 15407370
Change view
Click to view available options
Pulse Form:
AC Sine Wave, DC Square Wave
Unit Size:
Each
Product Code. Pulse Form unitSize
15407370 AC Sine Wave, DC Square Wave Each
1 options
Product Code. 15407370 Supplier BTX™ Supplier No. 452030
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
Allows for hands free operation of Gemini X2, ECM2001+, ECM830 and ECM630 generator (touch screen models). Harvard Apparatus Gemini X2 Electroporation Footswitch functions as the start button on front of the generator.
  • Allows for hands free operation of Gemini X2 generator
  • Desirable when conducting in vivo/in ovo gene delivery or nuclear transfer/cloning when both hands are needed for sample manipulation
  • Functions as start button on front of the generator
Specifications
For Use With (Application) Hands free operation of electroporation generators
For Use With (Equipment) Gemini X2, ECM 2001+, ECM 830, and ECM 630 Foot Pedal (compatible with touchscreen models)
Product Line BTX Electroporation
Product Type Electroporation accessory
Voltage 3000 VDC, 250 VAC
Description Gemini X2, ECM 2001+, ECM 830, and ECM 630 Foot Pedal
Quantity 1 Ea.
Sterility Non-sterile
Connection Type Electrode Connector Cable
Interface Footswitch
Pulse Form AC Sine Wave, DC Square Wave