missing translation for 'onlineSavingsMsg'
Learn More

BTX™ BTXPress™ Cytofusion Medium C

Product Code. 15427370
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15427370 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15427370 Supplier BTX™ Supplier No. 470001
  • ONLINE EXCLUSIVE PRICE
    594.00 €
    506.87 € / 500mL
    Save 87.13 €
    14% Off
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BTXPress™ Cytofusion Medium C is designed to promote efficiency while preserving high cell viability during cell fusion.

An advanced electrofusion buffer designed especially for hybridoma production. The low conductivity buffer is specially formulated to minimize cell turbulence during cell alignment and heating during electrofusion, for robust cell fusion efficiency and high cell viability. It is sterile-filtered from the highest quality non-animal, medical-grade reagents. It is the buffer of choice for many commercial biotech and pharmaceutical companies in their hybridoma generation process for monoclonal antibody discovery.

  • Optimized for eukaryotic electrofusion applications
  • Maximum fusion efficiency, high cell viability
  • Stable environment for cell alignment
  • Low conductivity for minimal heating
  • Physiological pH and balanced osmolarity
  • Contains no animal products

Specifications

Classification Electrofusion Buffer
Content And Storage 2°C to 8°C
Color Clear
Description BTXpress Cytofusion Medium C
Cell Type Non-animal
Sample Type Cells
Promoter None (Promoterless)
Transfection Technique Electroporation
For Use With (Equipment) BTX&trade ECM2001, Legacy ECM2001
For Use With (Application) Cell Fusion, Hybridoma Production
Includes 500 mL Buffer
pH 7.2 ± 0.2
Product Line BTX Electroporation/Electrofusion
Product Type Electrofusion Buffer
Quantity 500 mL
Reagent Type Medical-grade
Sterility Sterile
Volume (Metric) 500 mL
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.