missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ BTXPress™ Cytoporation Low Conductivity Medium T
Description
- Sterile-filtered from the highest quality non-animal, medical-grade reagents
- Buffer can be directly diluted in complete growth media for post-electroporation cell culture
- Optimized for eukaryotic electrofusion applications
- Maximum transfection efficiency, high cell viability
- Low conductivity for minimal heating
- Physiological pH and balanced osmolarity
- Contains no animal products
- Certified RNase and DNase free
Specifications
Specifications
| Format | Large volume electroporation |
| For Use With (Application) | Ex vivo or in vitro delivery of DNA, RNA, oligonucleotides, and siRNA |
| For Use With (Equipment) | BTX AgilePulse MAX Large Volume Electroporation System |
| Product Line | BTX Electroporation |
| Product Type | Electroporation Consumable |
| Description | BTXpress Cytoporation Low Conductivity Medium T |
| Life Span | 18 months from date of manufacture |
| Quantity | 500 mL |
| Sterility | Sterile |
| Content And Storage | 2°C to 8°C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?