missing translation for 'onlineSavingsMsg'
Learn More

BTX™ BTXPress™ Electroporation Buffer

Product Code. 15487340
Change view
Click to view available options
Quantity:
2 mL
5 mL
10 mL
No. of Reactions:
20
50
100
Unit Size:
10mL
2mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity No. of Reactions Unit Size
15487340 5 mL 50 5mL
15477340 2 mL 20 2mL
15417350 10 mL 100 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15487340 Supplier BTX™ Supplier No. 450802
  • ONLINE EXCLUSIVE PRICE
    514.00 €
    437.43 € / 5mL
    Save 76.57 €
    14% Off
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BTXPress™ Electroporation Buffer is a single buffer solution developed to quickly and efficiently deliver genes into mammalian cells that were previously considered hard to transfect by chemical and other non-viral methods.

BTXPress™ Electroporation Solution is the first electroporation reagent that meets all of your high performance transfection needs without sacrificing control over your experiment or your budget. It offers increased numbers of transfections per kit compared to other commercial alternatives, providing higher value to you. As a universal solution, the BTXPress™ Electroporation Solution can be used in other electroporators, including the Amaxa™, to deliver similar results without the typical high cost associated with these buffer kits. The BTXPress™ Electroporation Solution is offered as a kit including the BTX™ cuvettes Plus with transfer pipettes or as a buffer alone.

  • Efficient electroporation transfection
  • High efficiency transfection of difficult cell lines
  • High cell viability
  • Low toxicity resulting in improved cell viability
  • Single buffer for all cell types
  • One buffer used in place of standard electroporation buffers for all mammalian cell types

Specifications

Buffer Universal Electroporation Buffer
Content And Storage 2°C to 8°C
Color Clear
Description BTXpress Solution, 5.0 ml Bottle for up to 50 Reactions
Cell Type Mammalian
Format Bottle
Sample Type Cells
Promoter None (Promoterless)
Transfection Technique Electroporation
For Use With (Equipment) BTX™ and alternative electroporators
For Use With (Application) Transfection via Electroporation
Includes 5 mL Solution Only
Packaging Type Bottle
No. of Reactions 50
Product Line BTX Electroporation
Product Type Electroporation Buffer
Quantity 5 mL
Reagent Type Electroporation buffer
Sterility Sterile
Volume (Metric) Sample 5 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.