missing translation for 'onlineSavingsMsg'
Learn More

Notrax Bubble Sof-Tred™ Dyna-Shield™ Layer with Unique Bubble Design

Product Code. 17762108
Change view
Click to view available options
Length (Metric):
91 cm
150 cm
1830 cm
Width (Metric):
60 cm
91 cm
122 cm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Unit Size
17762108 150 cm 91 cm Each
17742108 91 cm 60 cm Each
17782098 1830 cm 60 cm Each
17702108 1830 cm 91 cm Each
17722108 1830 cm 122 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17762108 Supplier Notrax Supplier No. 417S0035BY
  • ONLINE EXCLUSIVE PRICE
    161.70 €
    139.19 € / Pack of 1
    Save 22.51 €
    13% Off
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Notrax Bubble Sof-Tred™ - Exceptional worker comfort due to 12.7 mm thick specially formulated compound. Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl.

  • Lengthwise corrugated rib pattern for sure footing
  • Exceptional worker comfort due to 127 mm thick specially formulated compound
  • Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl
  • Available with yellow borders along sides comply with OSHA code 1910-144
  • All 4 sides are bevelled and sealed to minimize tripping hazards

Specifications

Color Black/Yellow
Length (English) 59 in.
Length (Metric) 150 cm
Material Microcellular Vinyl
Type Industrial and Commercial Matting
Width (Metric) 91 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.