Learn More
Eppendorf™ Bucket for Rotor A-4-38
Product Code. 17677635 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
Rotor A-4-38
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17677635 Rotor A-4-38 Each
1 options
Product Code. 17677635 Supplier Eppendorf™ Supplier No. 5702764000
  • ONLINE EXCLUSIVE PRICE
    532.65 €
    483.00 € / Pack of 1
    Save 49.65 €
    9% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Bucket designed to hold 16 x 125 mm round bottom tubes.
  • For round bottom tubes
  • Hold 16 x 125 mm tubes
  • For rotor A-4-38
  • Accessory
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Rotor A-4-38
Product Type Adapters, Buckets and Accessories
Quantity 4/Pk.