missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Buckets or Lids for Thermo Scientific™ TX-150 Swinging Bucket Rotor

Product Code. 15145253 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Bucket
Bucket Lid
Capacity:
4 x 145 mL
8 x 50 mL
Not Applicable
Unit Size:
Each
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Capacity Unit Size
15145253 Bucket 8 x 50 mL Each
15165233 Bucket 4 x 145 mL Pack of 4
15152693 Bucket Lid Not Applicable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15145253 Supplier Thermo Scientific™ Supplier No. 75005703
  • ONLINE EXCLUSIVE PRICE
    1251.23 €
    999.00 € / Pack of 1
    Save 252.23 €
    20% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Round and conical buckets offered in sets of four

Round and conical buckets or lids offered in sets of four

TRUSTED_SUSTAINABILITY

Specifications

Capacity 8 x 50 mL
Product Type Bucket
Container Type Conical Bucket
Quantity Set of 4
Description Conical bucket, unsealed, no adapter needed
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.