missing translation for 'onlineSavingsMsg'
Learn More

Budesonide, TRC

Product Code. 17974445
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17974445 1 g 1g
17961905 100 mg 100mg
17921585 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17974445 Supplier Toronto Research Chemicals Supplier No. TRCB6894901G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51333-22-3
Molecular Formula C25 H34 O6
Molecular Weight (g/mol) 430.53
Synonym (11β,16α)-16,17-[Butylidenebis(oxy)]-11,21-dihydroxypregna-1,4-diene-3,20-dione, 2H-Naphth[2',1':4,5]indeno[1,2-d][1,3]dioxole, pregna-1,4-diene-3,20-dione deriv., 16α,17α-(Butylidenedioxy)-11β,21-dihydroxypregna-1,4-diene-3,20-dione, Barazone, Bidien, Budecort Aqua, Budenofalk, Budes, Budesomide, Budeson, Budesonide, Budesonide MMX, Cortivent, Entocort, Micronyl, Nebuampul, Novopulmon, Preferid, Pulmicort, Respules, Rhinocort, Rhinocort Aqua, Rhinocort respules, S 1320, Spirocort, Turbuhaler, Uceris, Uceris MMX
IUPAC Name 16α,17-[(1RS)-Butylidenebis(oxy)]-11β,21-dihydroxypregna-1,4-diene-3,20-dione
SMILES CCCC1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO

Specifications

Quantity 1 g
InChI Formula InChI=1S/C25H34O6/c1-4-5-21-30-20-11-17-16-7-6-14-10-15(27)8-9-23(14,2)22(16)18(28)12-24(17,3)25(20,31-21)19(29)13-26/h8-10,16-18,20-22,26,28H,4-7,11-13H2,1-3H3/t16-,17-,18-,20+,21?,22+,23-,24-,25+/m0/s1
Formula Weight 430.2355
Percent Purity >95%
Chemical Name or Material Budesonide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.