Learn More
Brand™ Counting Chambers, Burker-Turk pattern, BLAUBRAND™, double ruling
Product Code. 10333411 Shop All Brand Products
Change view
Click to view available options
Description:
Double-ruling, w/ Spring Clips
Double-ruling, w/o Spring Clips
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
10333411 Double-ruling, w/o Spring Clips Each
10297390 Double-ruling, w/ Spring Clips Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10333411 Supplier Brand™ Supplier No. 719505
  • ONLINE EXCLUSIVE PRICE
    64.90 €
    47.71 € / Pack of 1
    Save 17.19 €
    26% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
BLAUBRAND™ quality. Outstanding measurement precision. 100% certified quality.
  • Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.
  • Combination of the Burker and Thoma systems.The ruling shows 9 large squares of 1 mm2 each.
  • The large squares are subdivided into 16 group squares with 0.2 mm sides. in the central large square, each group square is subdivided into 16 mini squares with 0.05 mm sides ( 0.0025 mm2).
TRUSTED_SUSTAINABILITY
Specifications
Product Type Counting Device
Description Double-ruling, w/o Spring Clips
Quantity 1Each
Depth (Metric) 0.1mm