missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 10.0 (20°C), Violet colored, borax / sodium hydroxide, Honeywell Fluka™

Catalog Number 15634080
Click to view available options
Quantity:
500 mL
1 L
6 x 500 mL
10 L
6 x 1 L
This item is not returnable. View return policy
1336-21-6
H5NO
35.05
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
CHEBI:18219
azanium;hydroxide
N.O
This item is not returnable. View return policy
1336-21-6
H5NO
35.05
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
CHEBI:18219
azanium;hydroxide
N.O

Violet colored, borax / sodium hydroxide

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
pH 10.0
Quantity 500 mL
UN Number NONH for all modes of transport
Chemical Name or Material Buffer solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'