Learn More
Thermo Scientific™ Buffered Peptone Water (ISO) (Dehydrated)
Description
Pre-enrich food samples prior to the isolation of Salmonella species with this non-selective medium while adhering to ISO 11133:2014
Pre-enrich food samples prior to the isolation of Salmonella species using Thermo Scientific™ Oxoid™ Buffered Peptone Water (ISO) (Dehydrated). Buffered Peptone Water is a non-selective pre-enrichment medium and is tested in accordance with ISO 65791 and ISO 11133:20142. It also provides conditions for resuscitation of cells that have been injured by processes of food preservation.
- Pre-enrich food samples prior to isolation of Salmonella species
- Allow high resuscitation of sub-lethally injured cells with medium
- Low numbers and/or sub-lethally injured Salmonella spp. cells that may be in test food sample
- Supports high growth and recovery rate, rich in nutrients
- Prevents bacterial damage due to changes in medium pH
Specifications
Specifications
| Certifications/Compliance | ISO 6579:2002 |
| Form | Powder |
| Product Type | Buffered Peptone Water |
| For Use With (Application) | Allows cells time to repair and multiply before being introduced into a selective culture, Improving chances of recovering Salmonella species from the sample |
| Quantity | 5 kg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.