Learn More
Buffered Peptone Water Ready Bag
Description
Save time in preparing selective enrichment medium for the isolation of Salmonella spp. from food samples using this medium in ready prepared bags.
Save time in preparing selective enrichment medium for the isolation of Salmonella spp. from food samples using Thermo Scientific™ Oxoid™ Buffered Peptone Water Ready Bag. The autoclaved medium is supplied in a ready bag to suit your workflow. Medium can be dispensed through the outlet tube as required.
Shorten your workflow with easy-to-use Buffered Peptone Water Ready Bag. Ready to go when you are, the bags are highly adaptable to laboratory workflows and automated equipment, such as dilutors, with no further supplementation required.
- Cost-effective: no need for weighing of media or sterilization, reducing the burden of media preparation, which frees up resources and equipment.
- Easy to use: easy to dispense with minimum training
- Safe: eliminate handling of glassware, weighing of culture media, and the need to add supplementary reagents. Empty bags can be disposed of with normal laboratory waste
For added convenience, it is recommended that Ready Bag Enrichment Media are dispensed using the Thermo Scientific Automated Diluter (DB4150A).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Color | Straw, clear |
| Content And Storage | 2 to 25°C |
| Description | Buffered Peptone Water Ready Bag |
| Format | 3L bag with connection ports in a box |
| Product Type | Pre-enrichment Medium |
| Fill Weight | 3020mL ± 20mL |
| Quantity | 4/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.