missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Lysis Buffer LBP

Product Code. 15839127
Change view
Click to view available options
Quantity:
30 mL
125 mL
Chemical Name or Material:
Binding Solution BS
Lysis buffer LBP
Unit Size:
125mL
30mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Chemical Name or Material unitSize
15839127 125 mL Lysis buffer LBP 125mL
15849127 30 mL Binding Solution BS 30mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15839127 Supplier Macherey-Nagel™ Supplier No. 740906.125
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Additional buffer solutions for use with NucleoSpin™ RNA Plus Kits, for efficient and quick RNA isolation from cells and tissues.

NucleoSpin™ RNA Plus Kits

  • gDNA Removal Columns included – no time consuming rDNase digestion
  • Convenient handling – lysate clearing and gDNA removal using the same column (in one step)
  • Novel lysis buffer – exceptionally efficient, with no mercaptoethanol or TCEP necessary
  • Two column silica membrane system (one for DNA removal, one for RNA isolation)
  • Downstream applications: real-time RT-PCR, Northern blotting, primer extension, array technology, RNase protection assays

Specifications

Chemical Name or Material Lysis buffer LBP
CAS 593-84-0
Color Slightly Yellow
For Use With (Application) RNA isolation from cells and tissue
For Use With (Equipment) NucleoSpin RNA Plus kit
pH 6.5 to 7.5
Physical Form Liquid
Quantity 125 mL
Formulation Guanidinium thiocyanate; Water
Odor Odorless
compliance-icons
Product Identifier
  • Buffer LBP (125 mL)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contain: guanidium thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.