missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Comes complete with an elastic cord for dedicating that pipettor to a specific reagent bottle
- Tapered, silicone rubber seal provides an airtight fit on all size pipettes
- Can be opened, cleaned and steam autoclaved at 121°C (250°F)
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | Red |
| Material | Silicone Rubber |
| For Use With (Equipment) | Westergren or Wintrobe sedimentation tubes |
| Includes | Elastic Cord |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?