Learn More
Atago™ Bulbs
Product Code. 11741579
Change view
Click to view available options
Type:
Bulb
Unit Size:
Pack of 5
Product Code. Type unitSize
11741579 Bulb Pack of 5
1 options
Product Code. 11741579 Supplier Atago™ Supplier No. RE1170
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Use this bulb with Atago™ Illuminators for Abbe Refractometers.
  • Lamp for illuminator
  • 8V
  • 0.15A
  • Available in packs of five
Specifications
Type Bulb
For Use With (Equipment) Abbe Refractometers NAR-1T, NAR-2T, NAR-3T, NAR-4T
Description Lamp for illuminator (8V, 0.15A)
Quantity 5 Pk.