missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ULBP-2 His-tag Recombinant Protein

Product Code. 17986231
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17986231 1 mg 1mg
17925711 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17986231 Supplier Invitrogen™ Supplier No. RP88178
  • ONLINE EXCLUSIVE PRICE
    2360.00 €
    2100.40 € / 1mg
    Save 259.60 €
    11% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (900 μL) be added to the vial to prepare a stock solution of 0.25 mg/mL. Concentration is measured by UV.

ULBP2 encodes a major histocompatibility complex (MHC) class I-related molecule that binds to the NKG2D receptor on natural killer (NK) cells to trigger release of multiple cytokines and chemokines that in turn contribute to the recruitment and activation of NK cells. The encoded protein undergoes further processing to generate the mature protein that is either anchored to membrane via a glycosylphosphatidylinositol moiety, or secreted. Many malignant cells secrete the encoded protein to evade immunosurveillance by NK cells. This gene is located in a cluster of multiple MHC class I-related genes on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BZM5
For Use With (Application) Control, Neutralization
Format Lyophilized
Formulation PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 80328
Molecular Weight (g/mol) 23.18 kDa
Name Human ULBP-2 His-tag
pH Range 7.4
Purification Method Purified
Quantity 1 mg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias ALCAN-alpha; N2DL2; N2DL-2; NKG2D ligand 2; NKG2DL2; RAET1H; Retinoic acid early transcript 1H; UL16 binding protein 2; UL16-binding protein 2; ULBP2; UNQ463/PRO791
Common Name ULBP2
Gene Symbol ULBP2
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence Human ULBP2, amino acids Met1-Ser217 (Accession # Q9BZM5) with a C-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥97% as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.