missing translation for 'onlineSavingsMsg'
Learn More

Bumetanide 3-(Butyl(nitroso)amino), TRC

Product Code. 30137802
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137802 250 mg 250mg
30006707 100 mg 100mg
30004707 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137802 Supplier Toronto Research Chemicals Supplier No. TRCB989550250MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2490432-02-3
Molecular Formula C17H19N3O6S
Molecular Weight (g/mol) 393.41
Synonym 3-(Butyl(nitroso)amino)-4-phenoxy-5-sulfamoylbenzoic Acid
IUPAC Name 3-[butyl(nitroso)amino]-4-phenoxy-5-sulfamoyl-benzoic acid
SMILES CCCCN(N=O)c1cc(cc(c1Oc2ccccc2)S(=O)(=O)N)C(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C17H19N3O6S/c1-2-3-9-20(19-23)14-10-12(17(21)22)11-15(27(18,24)25)16(14)26-13-7-5-4-6-8-13/h4-8,10-11H,2-3,9H2,1H3,(H,21,22)(H2,18,24,25)
Formula Weight 393.0995
Percent Purity >95%
Chemical Name or Material Bumetanide 3-(Butyl(nitroso)amino)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.