missing translation for 'onlineSavingsMsg'
Learn More

Bumetanide, MP Biomedicals™

Product Code. 11474601 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474601 5 g 5g
11464601 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474601 Supplier MP Biomedicals Supplier No. 0215426305
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Bumetanide is used to probe the role of prostaglandin E2 in the regulation of chloride secretion across porcine endometrial epithelial cells.

  • A study of the effects of bumetanide application and subsequent sodium potassium chloride cotransporter (NKCC1) inhibition in cultured liver cells has indicated increased alpha-1-smooth muscle actin expression in 2-day-cultured hepatic stellate cells.
  • An investigation of lactacidosis treatment of suspended G6 glial cells with bumetanide included at 0.1 mM has shown diminished cell swelling due to lactacidosis.

Chemical Identifiers

CAS 28395-03-1
Molecular Formula C17H20N2O5S
Synonym bumetanidum

Specifications

Quantity 5 g
Melting Point 230 to 231°C
Physical Form Powder
Purity ≥99%
Molecular Weight (g/mol) 364.416
Chemical Name or Material Bumetanide

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.