missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Bupropion hydrochloride

Product Code. 30001402
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001402 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001402 Supplier R&D Systems™ Supplier No. 2831/50
  • ONLINE EXCLUSIVE PRICE
    242.48 €
    211.05 € / 50mg
    Save 31.43 €
    12% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Non-selective inhibitor of dopamine and noradrenalin transporters

Non-selective inhibitor of dopamine and noradrenalin transporters (Ki values are 1.4, 2.8 and 45 μM for NET, DAT and SERT transporters respectively). Also inhibits neuronal nicotinic acetylcholine receptors. Displays antidepressant activity and augments nicotine self-administration at low doses in vivo.
TRUSTED_SUSTAINABILITY

Specifications

CAS 31677-93-7
Quantity 50 mg
Product Line Tocris
Molecular Formula C13H19Cl2NO
Formula Weight 276.2
Purity >99%
Molecular Weight (g/mol) 276.201
Chemical Name or Material Bupropion hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.