missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Polyethylene Eye Rinsing Bottle

Product Code. 10240372 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
600 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10240372 600 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10240372 Supplier Buerkle™ Supplier No. 25072000
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Leakproof due to valve and dust protection cap

  • Quick and easy to use
  • First aid treatment for eye accidents
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 20.29 oz.
Capacity (Metric) 600 mL
Certifications/Compliance DIN
Color Transparent
Material Polyethylene
Mounting Portable
Product Type Eye Rinsing Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.