missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Busulfan, TRC
Specifications
Specifications
| Chemical Name or Material | Busulfan |
| CAS | 55-98-1 |
| Molecular Formula | C6 H14 O6 S2 |
| Quantity | 50 mg |
| Synonym | Busulfan, Butane-1,4-diyl di(methanesulfonate), 1,4-Butanediol, 1,4-dimethanesulfonate (ACI), 1,4-Butanediol, dimethanesulfonate (8CI, 9CI), Methanesulfonic acid, tetramethylene ester (6CI), 1,4-Bis(methanesulfonoxy)butane, 1,4-Bis(methanesulfonyloxy)butane, 1,4-Butanediol dimesulfonate, 1,4-Butanediol dimesylate, 1,4-Dibutanediol dimethanesulfonate, 1,4-Dimethanesulfonoxybutane, 1,4-Dimethylsulfonyloxybutane, 2041CB, 4-Methylsulfonyloxybutyl methanesulfonate, AN 33501, Busilvex, Busulfex, Busulphan, Butane-1,4-diyl bis(methanesulfonate), CB 2041, Citosulfan, Glyzophrol, GT 2041, GT 41, Leucosulfan, Mablin, MeSH ID: D002066, Mielevcin, Mielosan, Mielucin, Milecitan, Mileran, Misulban, Mitostan, Myeloleukon, Myelosan, Mylecytan, Myleran, Myran, NCI C01592, NSC 750, Sulfabutin, Sulphabutin, Tetramethylene bis[methanesulfonate], X 149 |
| InChI Formula | InChI=1S/C6H14O6S2/c1-13(7,8)11-5-3-4-6-12-14(2,9)10/h3-6H2,1-2H3 |
| SMILES | CS(=O)(=O)OCCCCOS(=O)(=O)C |
| IUPAC Name | 4-methylsulfonyloxybutyl methanesulfonate |
| Molecular Weight (g/mol) | 246.3 |
| Formula Weight | 246.0232 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?