missing translation for 'onlineSavingsMsg'
Learn More

Butadiene monoxide, 98%

Product Code. 11399953
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399953 1 g 1g
11309963 5 g 5g
11319963 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399953 Supplier Thermo Scientific Alfa Aesar Supplier No. L00238.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 930-22-3 | C4H6O | 70.091 g/mol

Butadiene monoxide is used as key starting material for the synthesis of (-)-bulgecinine and (+)-broussonetine G.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butadiene monoxide is used as key starting material for the synthesis of (-)-bulgecinine and (+)-broussonetine G.

Solubility
Miscible with ethanol, ethyl ether, benzene and organic solvents.

Notes
Moisture sensitive. Store in cool place. Incompatible with acids, bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-22-3
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00005149
InChI Key GXBYFVGCMPJVJX-UHFFFAOYSA-N
Synonym butadiene monoxide, oxirane, ethenyl, 3,4-epoxy-1-butene, 2-vinyloxirane, butadiene epoxide, 1,2-epoxy-3-butene, vinyloxirane, butadiene oxide, butadiene monoepoxide, butadiene monooxide
PubChem CID 13586
IUPAC Name 2-ethenyloxirane
SMILES C=CC1CO1

Specifications

Melting Point -135°C
Density 0.87
Boiling Point 65°C to 66°C
Flash Point −50°C (−58°F)
Odor Recognizable
Refractive Index 1.418
Quantity 1 g
UN Number UN1993
Beilstein 103170
Sensitivity Moisture sensitive
Solubility Information Miscible with ethanol,ethyl ether,benzene and organic solvents.
Formula Weight 70.09
Percent Purity 98%
Chemical Name or Material Butadiene monoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.