missing translation for 'onlineSavingsMsg'
Learn More

Butamirate Citrate, TRC

Product Code. 30011407
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011407 10 mg 10mg
30010908 50 mg 50mg
30010939 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011407 Supplier Toronto Research Chemicals Supplier No. TRCB69002510MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18109-81-4
Molecular Formula C18 H29 N O3 . C6 H8 O7
Molecular Weight (g/mol) 499.55
Synonym Benzeneacetic acid, α-ethyl-, 2-[2-(diethylamino)ethoxy]ethyl ester, 2-hydroxy-1,2,3-propanetricarboxylate (1:1), Butyric acid, 2-phenyl-, 2-[2-(diethylamino)ethoxy]ethyl ester, citrate (1:1) (8CI), Ethanol, 2-[2-(diethylamino)ethoxy]-, 2-phenylbutyrate (ester), citrate (1:1) (8CI), 2-[2-(Diethylamino)ethoxy]ethyl-2-phenylbutyrate citrate (1:1), Abbott 36581, Acodeen, Butamirate citrate, Cough Cut, HH 197, Panatus, Sincodeen, Sincodin, Sincodix, Sinecod, Sinecond
IUPAC Name 2-[2-(diethylamino)ethoxy]ethyl 2-phenylbutanoate;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES CCC(C(=O)OCCOCCN(CC)CC)c1ccccc1.OC(=O)CC(O)(CC(=O)O)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H29NO3.C6H8O7/c1-4-17(16-10-8-7-9-11-16)18(20)22-15-14-21-13-12-19(5-2)6-3;7-3(8)1-6(13,5(11)12)2-4(9)10/h7-11,17H,4-6,12-15H2,1-3H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)
Formula Weight 499.2417
Percent Purity >95%
Chemical Name or Material Butamirate Citrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.