missing translation for 'onlineSavingsMsg'
Learn More

Butanal, TRC

Product Code. 30333843
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30333843 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30333843 Supplier Toronto Research Chemicals Supplier No. TRCB6898901G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Butanal
CAS 123-72-8
Molecular Formula C4 H8 O
Quantity 1 g
Synonym Butyraldehyde (8CI), Butal, Butaldehyde, Butanaldehyde, Butyl aldehyde, Butyral, Butyric aldehyde, Butyrylaldehyde, NSC 62779, n-Butanal, n-Butyl aldehyde, n-Butyraldehyde, n-Butyric aldehyde
InChI Formula InChI=1S/C4H8O/c1-2-3-4-5/h4H,2-3H2,1H3
SMILES CCCC=O
IUPAC Name butanal
Molecular Weight (g/mol) 72.11
Formula Weight 72.0575
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.