missing translation for 'onlineSavingsMsg'
Learn More

Buthiazide, TRC

Product Code. 30010105
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010105 50 mg 50mg
30009197 25 mg 25mg
30006063 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010105 Supplier Toronto Research Chemicals Supplier No. TRCB69003550MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2043-38-1
Molecular Formula C11 H16 Cl N3 O4 S2
Molecular Weight (g/mol) 353.85
Synonym 2H-1,2,4-Benzothiadiazine-7-sulfonamide, 6-chloro-3,4-dihydro-3-(2-methylpropyl)-, 1,1-dioxide, 2H-1,2,4-Benzothiadiazine-7-sulfonamide, 6-chloro-3,4-dihydro-3-isobutyl-, 1,1-dioxide (6CI,7CI,8CI), 3-Isobutyl-6-chloro-3,4-dihydro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide, 3500S, Buthiazide, Butizid, Butizide, Eunephran, Isobutylhydrochlorothiazide, Modenol, S 3500, Saltucin, Su 6187, Thiabutazide
IUPAC Name 6-chloro-3-(2-methylpropyl)-1,1-dioxo-3,4-dihydro-2H-1λ^{6},2,4-benzothiadiazine-7-sulfonamide
SMILES CC(C)CC1Nc2cc(Cl)c(cc2S(=O)(=O)N1)S(=O)(=O)N

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H16ClN3O4S2/c1-6(2)3-11-14-8-4-7(12)9(20(13,16)17)5-10(8)21(18,19)15-11/h4-6,11,14-15H,3H2,1-2H3,(H2,13,16,17)
Formula Weight 353.0271
Percent Purity >95%
Chemical Name or Material Buthiazide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.