missing translation for 'onlineSavingsMsg'
Learn More

Butyl acetate, CHROMASOLV™ Plus, for HPLC, 99.7%, Solstice

Product Code. 15622870
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
Packaging:
Glass Bottle
Unit Size:
1 L
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15622870 6 x 1 L Glass Bottle Pack of 6
15612870 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15622870 Supplier Honeywell-Chromasolv Supplier No. 2706876X1L
  • ONLINE EXCLUSIVE PRICE
    551.00 €
    452.55 € / Pack of 6
    Save 98.45 €
    17% Off
View all promotions
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHROMASOLV™ Plus, for HPLC, 99.7%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Absorbance max. 0.01 at 400 nm, max. 0.01 at 320 nm, max. 0.02 at 300 nm, max. 0.04 at 275 nm, max. 0.20 at 260 nm
CAS Max % 100.0000%
Melting Point -78°C (literature)
Density 0.88 g/mL at 25 °C(lit.)
Boiling Point 126.2°C
Flash Point 27°C
Packaging Glass Bottle
Linear Formula CH3COO(CH2)3CH3
Quantity 6 x 1 L
UN Number UN1123
Beilstein 1741921
Formula Weight 116.16
Percent Purity >99.65%
Grade HPLC
Chemical Name or Material Butyl acetate, CHROMASOLV™ Plus, for HPLC, 99.7%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.