missing translation for 'onlineSavingsMsg'
Learn More

Butyl acrylate, 99+%, stabilized

Product Code. 10308533 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10308533 10 L Plastic drum 10 L
10667182 1 L Glass Bottle 1 L
10751491 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10308533 Supplier Thermo Scientific Acros Supplier No. 215860100
  • ONLINE EXCLUSIVE PRICE
    380.00 €
    368.00 € / 10 L
    Save 12.00 €
    3% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 141-32-2 | C7H12O2 | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-32-2 , 150-76-5
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00009446
InChI Key CQEYYJKEWSMYFG-UHFFFAOYSA-N
Synonym butyl acrylate, n-butyl acrylate, acrylic acid butyl ester, n-butyl propenoate, 2-propenoic acid, butyl ester, butyl 2-propenoate, butylacrylate, acrylic acid, butyl ester, acrylic acid n-butyl ester, butylester kyseliny akrylove
PubChem CID 8846
ChEBI CHEBI:3245
IUPAC Name butyl prop-2-enoate
SMILES CCCCOC(=O)C=C

Specifications

Acidity 0.01% max. (as acrylic acid)
Melting Point -64.0°C
Density 0.8900g/mL
Boiling Point 145.0°C
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic drum
Linear Formula H2C=CHCO2(CH2)3CH3
Refractive Index 1.4170 to 1.4190
Quantity 10 L
Beilstein 02, II, 388
Merck Index 15, 1541
Specific Gravity 0.89
Solubility Information Solubility in water: 1.4g/L (20°C)
Stabilizer 10 to 20ppm MEHQ
Viscosity 0.869 mPa.s (20°C)
Formula Weight 128.17
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Butyl acrylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.