missing translation for 'onlineSavingsMsg'
Learn More

Butyl Acrylate, TRC

Product Code. 30342368
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30342368 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342368 Supplier Toronto Research Chemicals Supplier No. TRCB69323025ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acrylic acid butyl ester
CAS 141-32-2
Molecular Formula C7 H12 O2
Quantity 25 mL
Synonym Acrylic acid butyl ester (6CI,8CI), 2-Propenoic acid butyl ester, Acrylic acid n-butyl ester, BA 100PPM, Butyl 2-propenoate, Butyl acrylate, Butyl propenoate, DL 325, EM 24, NSC 5163, n-Butyl acrylate
InChI Formula InChI=1S/C7H12O2/c1-3-5-6-9-7(8)4-2/h4H,2-3,5-6H2,1H3
SMILES CCCCOC(=O)C=C
IUPAC Name butyl prop-2-enoate
Molecular Weight (g/mol) 128.17
Formula Weight 128.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.