missing translation for 'onlineSavingsMsg'
Learn More

Butyl chloroformate, 98%

Product Code. 10532781 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10532781 25 mL Glass bottle 25mL
10144150 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10532781 Supplier Thermo Scientific Supplier No. 180170250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 592-34-7 | C5H9ClO2 | 136.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-34-7
MDL Number MFCD00000650
InChI Key NRDQFWXVTPZZAZ-UHFFFAOYSA-N
Synonym butyl chloroformate, n-butyl chloroformate, butylchloroformate, carbonochloridic acid, butyl ester, chloroformic acid butyl ester, butyl chlorocarbonate, n-butyl chlorocarbonate, formic acid, chloro-, butyl ester, butyl-chloroformate, butylchloro-formate
PubChem CID 61140
IUPAC Name butyl carbonochloridate
SMILES CCCCOC(=O)Cl

Specifications

Melting Point -70.0°C
Density 1.0600g/mL
Boiling Point 142.0°C
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C5H9ClO2
Linear Formula ClCO2(CH2)3CH3
Refractive Index 1.4110 to 1.4130
Quantity 25 mL
Beilstein 03, II, 11
Specific Gravity 1.06
Solubility Information Solubility in water: decomposes in water. Other solubilities: soluble in butanol,ether,benzol,soluble in most common organic solvents
Molecular Weight (g/mol) 136.58
Viscosity 0.95 mPa.s (20°C)
Formula Weight 136.58
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Butyl chloroformate, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Butyl chloroformate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.