missing translation for 'onlineSavingsMsg'
Learn More

Butyl Digol, Extra Pure, SLR, Fisher Chemical™

Product Code. 10315220 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10315220 2.5 L 2.5 L
10040070 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10315220 Supplier Fisher Chemical Supplier No. B/5300/17
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 112-34-5
Molecular Formula C8H18O3
MDL Number 2881

Specifications

Melting Point -68°C
Color Colorless
Boiling Point 231°C
Packaging Amber glass bottle
Quantity 2.5 L
Formula Weight 162.23g/mol
Physical Form Liquid
compliance-icons
Product Identifier
  • 2-(2-Butoxyethoxy)ethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.