missing translation for 'onlineSavingsMsg'
Learn More

Butyl formate, 97%

Product Code. 10751822 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10751822 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10751822 Supplier Thermo Scientific Acros N. del fornitore 403541000
 Restricted Item
In Stock
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 592-84-7 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifica

Acidity 2mg KOH/g max.
CAS 592-84-7
Melting Point -91.0°C
Color Colorless to Yellow
Density 0.8940g/mL
Boiling Point 107.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C5H10O2
Linear Formula HCOO(CH2)3CH3
Refractive Index 1.3886 to 1.3906
MDL Number MFCD00003296
Quantity 100 g
Specific Gravity 0.894
Synonym n-butyl formate, formic acid, butyl ester, butyl methanoate, n-butyl methanoate, formic acid butyl ester, n-butylformate, butylester kyseliny mravenci, fema no. 2196, unii-0y9mz7vm9p, butylester kyseliny mravenci czech
Solubility Information Solubility in water: slightly soluble in water. Other solubilities: miscible with alcohols,ethers,oils,hydrocarbons,soluble in acetone,soluble in many organic solvents
InChI Key NMJJFJNHVMGPGM-UHFFFAOYSA-N
SMILES CCCCOC=O
IUPAC Name butyl formate
Molecular Weight (g/mol) 102.13
PubChem CID 11614
Viscosity <30 mPa.s (20°C)
Formula Weight 102.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Butyl formate, 98%
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons
Identificativo prodotto
  • Butyl formate
Avvertenza
  • Danger
Categoria di pericolo
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Consigli di prudenza
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.