missing translation for 'onlineSavingsMsg'
Learn More

Butyl methacrylate, 99%, stabilized

Product Code. 10616214 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
5 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10616214 5 L Plastic drum 5 L
10318640 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10616214 Supplier Thermo Scientific Supplier No. 216620051
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 97-88-1 | C8H14O2 | 142.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-88-1 , 150-76-5
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00009444
InChI Key SOGAXMICEFXMKE-UHFFFAOYSA-N
Synonym butyl methacrylate, n-butyl methacrylate, butylmethacrylate, 2-propenoic acid, 2-methyl-, butyl ester, butyl 2-methacrylate, butylmethacrylaat, 2-methyl-butylacrylat, butil metacrilato, 2-methyl-butylacrylaat, 2-methyl-butylacrylate
PubChem CID 7354
IUPAC Name butyl 2-methylprop-2-enoate
SMILES CCCCOC(=O)C(C)=C

Specifications

Melting Point -75.0°C
Density 0.8900g/mL
Boiling Point 160.0°C to 163.0°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Linear Formula H2C=C(CH3)CO2(CH2)3CH3
Refractive Index 1.4225 to 1.4245
Quantity 5 L
Beilstein 02, III, 1286
Specific Gravity 0.89
Solubility Information Solubility in water: 3g/L in water (20°C). Other solubilities: very soluble in ether and ethanol,miscible in ethyl acetate
Stabilizer 10 to 110ppm MEHQ
Viscosity 1.1 mPa.s (20°C)
Formula Weight 142.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Butyl methacrylate, Stabilized
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.