missing translation for 'onlineSavingsMsg'
Learn More

Butyl Nitrite, TRC

Product Code. 30011705
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011705 5 g 5g
30005452 100 g 100g
30011578 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011705 Supplier Toronto Research Chemicals Supplier No. TRCB6939205G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-16-1
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
Synonym NSC 8426, n-Butyl Nitrite, Nitrous Acid, Butyl Ester
IUPAC Name butyl nitrite
SMILES O=NOCCCC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C4H9NO2/c1-2-3-4-7-5-6/h2-4H2,1H3
Formula Weight 103.0633
Chemical Name or Material Butyl Nitrite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.