missing translation for 'onlineSavingsMsg'
Learn More

Butyl Phenyl Phosphate, TRC

Product Code. 30137786
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137786 1 g 1g
30008007 500 mg 500mg
30008344 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137786 Supplier Toronto Research Chemicals Supplier No. TRCB8095901G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2752-95-6
Molecular Formula C16 H19 O4 P
Molecular Weight (g/mol) 306.29
Synonym Butyl phenyl phosphate
IUPAC Name butyl diphenyl phosphate
SMILES CCCCOP(=O)(Oc1ccccc1)Oc2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H19O4P/c1-2-3-14-18-21(17,19-15-10-6-4-7-11-15)20-16-12-8-5-9-13-16/h4-13H,2-3,14H2,1H3
Formula Weight 306.1021
Percent Purity >95%
Chemical Name or Material Butyl diphenyl phosphate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.