missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Butyl Sepharose™ 4 Fast Flow Chromatography Media

Product Code. 10317689 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
200 mL
Unit Size:
25mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10317689 25 mL 25mL
10155917 200 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10317689 Supplier Cytiva Supplier No. 17098010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for capture and intermediate purification of larger proteins

  • Based on Sepharose Fast Flow base matrix derivatized via uncharged, chemically-stable ether linkages
  • Hydrophobic interaction chromatography (HIC) media composed of cross-linked 4% agarose
  • Aliphatic butyl ligand immobilized to base matrix with ether linkage
  • Fast Flow matrix gives the adsorbent high chemical, physical and thermal stabilities
  • Dynamic capacity at low salt concentration
  • Used with all aqueous buffers commonly used in chromatography
  • Autoclavable at 121°C for 20 minutes in water
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mL
For Use With (Application) For Capture and Intermediate Purification of Larger Proteins
Product Type Butyl Sepharose 4 Fast Flow Media
Matrix Based on cross-linked 4% agarose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.