missing translation for 'onlineSavingsMsg'
Learn More

Butylated Hydroxyanisole, TRC

Product Code. 17941175
Change view
Click to view available options
Quantity:
1 g
5 g
100 g
Unit Size:
100g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17941175 1 g 1g
30005467 100 g 100g
30003728 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17941175 Supplier Toronto Research Chemicals Supplier No. TRCB6907551G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25013-16-5
Molecular Formula 2 C11 H16 O2
Molecular Weight (g/mol) 360.49
Synonym Phenol, tert-butyl-4-methoxy- (7CI,8CI), (1,1-Dimethylethyl)-4-methoxyphenol, 2(3)-tert-Butyl-4-hydroxyanisole, Antioxyne B, BHA, BHA (antioxidant), BOA, BOA (antioxidant), Butylated hydroxyanisole, Butylhydroxyanisole, E 320, Embanox, Grindox BHA, Protex, Sustane 1F, Tenox BHA, tert-Butyl-4-hydroxyanisole, tert-Butyl-4-methoxyphenol, tert-Butyl-p-Hydroxyanisol, tert-Butyl-p-hydroxyanisole, tert-Butylhydroxyanisole
IUPAC Name 2-tert-butyl-4-methoxyphenol;3-tert-butyl-4-methoxyphenol
SMILES COc1ccc(O)c(c1)C(C)(C)C.COc2ccc(O)cc2C(C)(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/2C11H16O2/c1-11(2,3)9-7-8(13-4)5-6-10(9)12;1-11(2,3)9-7-8(12)5-6-10(9)13-4/h2*5-7,12H,1-4H3
Formula Weight 360.2301
Chemical Name or Material tert-Butyl-4-hydroxyanisole (mixture of 2- and 3-isomer)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.