missing translation for 'onlineSavingsMsg'
Learn More

Butyltin Trichloride, TRC

Product Code. 30337862
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30337862 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337862 Supplier Toronto Research Chemicals Supplier No. TRCB80952025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material n-Butyl-tin trichloride
CAS 1118-46-3
Molecular Formula C4 H9 Cl3 Sn
Quantity 25 g
Synonym Butyltin trichloride (6CI), Butyltrichlorostannane, Butyltin trichloride (BuSnCl3), Butyltrichlorotin, Certincoat TC 100, Monobutyltin trichloride, NSC 67013, Trichlorobutylstannane, Trichlorobutyltin, n-Butyltin trichloride
InChI Formula InChI=1S/C4H9.3ClH.Sn/c1-3-4-2;;;;/h1,3-4H2,2H3;3*1H;/q;;;;+3/p-3
SMILES CCCC[Sn](Cl)(Cl)Cl
IUPAC Name butyl(trichloro)stannane
Molecular Weight (g/mol) 282.18
Formula Weight 281.8792
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.