missing translation for 'onlineSavingsMsg'
Learn More

Butyraldehyde, ≥99.0%, Honeywell™

Product Code. 15605810 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15605810 100 mL Glass bottle 100mL
15615810 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15605810 Supplier Honeywell Research Chemicals Supplier No. 538191100ML
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

≥99.0%, dry

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-72-8
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00007023
InChI Key ZTQSAGDEMFDKMZ-UHFFFAOYSA-N
Synonym butyraldehyde, n-butyraldehyde, butyral, 1-butanal, butyric aldehyde, butanaldehyde, butaldehyde, butalyde, butal, n-butanal
PubChem CID 261
ChEBI CHEBI:15743
IUPAC Name butanal
SMILES CCCC=O

Specifikationer

Melting Point -81°C
Color Colorless
Density 0.8g/mL at 25°C
Boiling Point 46°C to 50°C
Packaging Glass bottle
Linear Formula CH3CH2CH2CHO
Quantity 100 mL
UN Number UN1129
Beilstein 506061
Formula Weight 72.11g/mol
Percent Purity ≥99.0%
Chemical Name or Material Butyraldehyde
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.