missing translation for 'onlineSavingsMsg'
Learn More

Butyraldehyde, 99%

Product Code. 10686672 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10686672 1 L Glass bottle 1 L
10540001 100 mL Glass bottle 100mL
10512094 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10686672 Supplier Thermo Scientific Acros Supplier No. 108090010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 123-72-8 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-72-8
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00007023
InChI Key ZTQSAGDEMFDKMZ-UHFFFAOYSA-N
Synonym butyraldehyde, n-butyraldehyde, butyral, 1-butanal, butyric aldehyde, butanaldehyde, butaldehyde, butalyde, butal, n-butanal
PubChem CID 261
ChEBI CHEBI:15743
IUPAC Name butanal
SMILES CCCC=O

Specifications

Melting Point -96.0°C
Color Colorless
Density 0.8170g/mL
Boiling Point 75.0°C
Flash Point −12°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2CHO
Refractive Index 1.3770 to 1.3830
Quantity 1 L
Beilstein 01, 662
Merck Index 15, 1595
Specific Gravity 0.817
Solubility Information Solubility in water: 7.1% (25°C). Other solubilities: miscible with ethanol, ether, ethyl acetate,, acetone and toluene
Viscosity 0.43 mPa.s (20°C)
Formula Weight 72.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Butyraldehyde
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Butyraldehyde
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.