missing translation for 'onlineSavingsMsg'
Learn More

Butyrolactone 3, TRC

Product Code. 30008464
Change view
Click to view available options
Quantity:
1 mg
5 mg
50 mg
Unit Size:
1mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008464 5 mg 5mg
30009303 1 mg 1mg
30011480 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008464 Supplier Toronto Research Chemicals Supplier No. TRCB7610105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 778649-18-6
Molecular Formula C9 H12 O4
Molecular Weight (g/mol) 184.189
Synonym (2R,3S)-rel-Tetrahydro-4-methylene-5-oxo-2-propyl-3-furancarboxylic Acid, 4-Methylene-5-oxo-2-propyl-tetrahydrofuran-3-carboxylic Acid, 3-Furancarboxylic acid, tetrahydro-4-methylene-5-oxo-2-propyl-, (2R,3S)-rel-, rel-(2R,3S)-Tetrahydro-4-methylene-5-oxo-2-propyl-3-furancarboxylic acid, 4-Methylene-5-oxo-2-propyl-tetrahydrofuran-3-carboxylic acid, Butyrolactone 3, MB 3
IUPAC Name (2S,3R)-4-methylene-5-oxo-2-propyl-tetrahydrofuran-3-carboxylic acid
SMILES CCC[C@@H]1OC(=O)C(=C)[C@H]1C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C9H12O4/c1-3-4-6-7(8(10)11)5(2)9(12)13-6/h6-7H,2-4H2,1H3,(H,10,11)/t6-,7+/m0/s1
Formula Weight 184.0736
Chemical Name or Material Butyrolactone 3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.