missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyrophenone-2',3',4',5',6'-d5, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Butyrophenone-2',3',4',5',6'-D5 |
| CAS | 39058-44-1 |
| Molecular Formula | C10 D5 H7 O |
| Quantity | 0.5 g |
| Synonym | 1-Butanone, 1-(phenyl-d5)- (9 CI), Butyrophenone-2',3',4',5',6'-d5, 1-(Phenyl-d5)1-butanone, Butyrophenone-2',3',4',5',6'-D5, Butyrophenone-d5 |
| InChI Formula | InChI=1 S/C10H12O/c1-2-6-10(11)9-7-4-3-5-8-9/h3-5,7-8 H,2,6H2,1H3/i3D,4 D,5 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)CCC)c([2 H])c1[2 H] |
| IUPAC Name | 1-(2,3,4,5,6-pentadeuteriophenyl)butan-1-one |
| Molecular Weight (g/mol) | 153.2325 |
| Formula Weight | 153.1202 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction