Learn More
3M™ BX™ Spectacles
Product Code. 10833241 Shop All 3M Products
Change view
Click to view available options
Style: +1.50 M
Product Code.
10833241 Style: +1.50 M
1 options
Product Code. 10833241 Supplier 3M™ Supplier No. 7000043082
Quantity Price
1 14.31 € / Pack of 1
20 12.84 € / Pack of 1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
A sleek, contoured and fully adjustable model designed for today's flexible work force. 3M™ BX™ Spectacles are also available in bi-focal lenses that provide magnification for reading or precision work.
  • Soft, universal-fit nose bridge
  • Soft tip temples to improve comfort
  • Excellent protection against flying particles
  • Special anti-fog coating reduces fogging and ensures clearer vision
  • Adjustable length temples with pantoscopic system (3 angles for custom fit)
TRUSTED_SUSTAINABILITY
Specifications
Lens Tint Clear
Product Type Safety Glass 1.5 M
No. per Pack 1 Ea.
Lens Material Polycarbonate