Learn More
Thermo Scientific™ Bz-rA Phosphoramidite, standard grade, serum vial bottle
Shop All Thermo Scientific ProductsBeschreibung
Bz-rA CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with Benzoyl (Bz) base protection, for use in oligonucleotide synthesis. RNA phosphoramidites contain a 2'-OH protecting group, tBDMS (tert-butyl dimethylsilyl). Each base has unique protecting groups designed specifically to protect the RNA during the chemical synthesis process, and produce the highest amount of full length product. Alternate base protection options are also available to allow for faster deprotection and longer oligonucleotide synthesis. The 'Standard Grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 60 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.
Features:
• Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-N6-benzoyl-2'-O-TBDMSi-adenosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula: C53H66N7O8PSi
• Base: rA
• Base Protection: Bz
• Formula weight: 988.2 g/mol
• CAS number: 104992-55-4
• HPLC Purity: ≥98.0%
• 31P NMR Purity: ≥98.0%; Sum of non-primary peaks in 140-152 ppm range: ≤0.5%
Spezifikation
Spezifikation
| For Use With (Equipment) | AKTA Synthesizers, Applied Biosystems Synthesizers |
| Product Type | Bz-rA CEP |
| Content And Storage | Store in freezer (at or below -20°C). |
| Nucleoside Type | Adenosine (A) |
| Shipping Condition | Room Temperature |
| Purity | 98% |
| Quantity | 2 g |
| Reagent Type | Phosphoramidite |
| Grade | Standard |
| Nucleic Acid Type | RNA |
| Mehr anzeigen |
For Research Use Only. Not for use in diagnostic procedures.
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.